2,4-Dimethoxytoluene ≥97%

Supplier: AOBChem USA

Warning

32419-25G 32419-5G
76902-292EA 375.56 USD
76902-292 76902-294
2,4-Dimethoxytoluene ≥97%
2,4-Dimethoxytoluene

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00010749
CAS Number: 38064-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR