2,6-Dichloro-4-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32403-1G 32403-25G 32403-500MG 32403-5G
76902-278EA 185.66 USD
76902-278 76902-280 76902-282 76902-284
2,6-Dichloro-4-methylphenylboronic acid pinacol ester ≥97%
2,6-Dichloro-4-methylphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BCl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD28142032
CAS Number: 2121511-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR