2,5-Dimethoxy-4-formylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (4-Formyl-2,5-dimethoxyphenyl)boronic acid

32347-1G 32347-25G 32347-500MG 32347-5G
76902-266EA 155.96 USD
76902-266 76902-268 76902-270 76902-272
2,5-Dimethoxy-4-formylphenylboronic acid ≥97% research grade
2,5-Dimethoxy-4-formylphenylboronic acid
Formula: C₉H₁₁BO₅
Storage Temperature: Refrigerator
MDL Number: MFCD32200303
CAS Number: 2468045-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR