2-Bromo-3-chloropyridin-4-ol ≥97%

Supplier: AOBChem USA

Warning

32269-1G 32269-25G 32269-500MG 32269-5G
76902-252EA 292.81 USD
76902-252 76902-254 76902-256 76902-258
2-Bromo-3-chloropyridin-4-ol ≥97%
2-Bromo-3-chloropyridin-4-ol
Formula: C₅H₃BrClNO
MW: 208.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18257590
CAS Number: 1211525-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR