2-Chloro-4-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32251-1G 32251-25G 32251-500MG 32251-5G
76902-242EA 120.95 USD
76902-242 76902-244 76902-246 76902-248
2-Chloro-4-formylbenzoic acid ≥97%
2-Chloro-4-formylbenzoic acid
Formula: C₈H₅ClO₃
MW: 184.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11110998
CAS Number: 1289063-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR