2,6-Difluoro-3-hydroxymethylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32221-1G 32221-25G 32221-500MG 32221-5G
76902-228EA 147.47 USD
76902-228 76902-230 76902-232 76902-234
2,6-Difluoro-3-hydroxymethylphenylboronic acid pinacol ester ≥97%
2,6-Difluoro-3-hydroxymethylphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30344689
CAS Number: 2121511-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR