2,6-Difluoro-3-formylbenzoic acid 95%

Supplier: AOBChem USA

Warning

32218-1G 32218-25G 32218-5G 32218-500MG
76902-214EA 106.09 USD
76902-214 76902-216 76902-218 77646-036
2,6-Difluoro-3-formylbenzoic acid 95%
2,6-Difluoro-3-formylbenzoic acid
Formula: C₈H₄F₂O₃
MW: 186.112 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26687989
CAS Number: 1500192-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR