2-Bromo-3-chloro-6-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32123-1G 32123-25G 32123-5G
76902-188EA 117.76 USD
76902-188 76902-190 76902-192
2-Bromo-3-chloro-6-fluorobenzyl alcohol ≥97%
2-Bromo-3-chloro-6-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28141932
CAS Number: 1823576-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR