2-Chloro-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32030-1G 32030-25G 32030-5G
76902-154EA 106.09 USD
76902-154 76902-156 76902-158
2-Chloro-4-isopropoxybenzoic acid ≥97%
2-Chloro-4-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD17256599
CAS Number: 334018-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR