3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoro-4-hydroxyphenylboronic acid pinacol ester

31915-1G 31915-25G 31915-500MG 31915-5G
76902-130EA 183.54 USD
76902-130 76902-132 76902-134 76902-136
3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%
3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Formula: C₁₂H₁₅BF₂O₃
MW: 256.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414561
CAS Number: 1029439-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR