2-Methoxy-3-nitro-5-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester, 2-Methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

31900-1G 31900-25G 31900-500MG 31900-5G
76902-116EA 180.36 USD
76902-116 76902-118 76902-120 76902-122
2-Methoxy-3-nitro-5-pyridineboronic acid pinacol ester ≥97%
2-Methoxy-3-nitro-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BN₂O₅
Storage Temperature: Ambient
MDL Number: MFCD12923397
CAS Number: 1083168-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR