2,4-Difluoro-5-ethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

31778-25G 31778-5G
76902-088EA 453.01 USD
76902-088 76902-090
2,4-Difluoro-5-ethoxyphenylboronic acid ≥97%
2,4-Difluoro-5-ethoxyphenylboronic acid
Formula: C₈H₉BF₂O₃
MW: 201.963 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783133
CAS Number: 900175-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR