2,4-Difluoro-5-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

31725-1G 31725-25G 31725-500MG 31725-5G
76902-046EA 165.51 USD
76902-046 76902-048 76902-050 76902-052
2,4-Difluoro-5-methoxyphenylboronic acid ≥97%
2,4-Difluoro-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD21609510
CAS Number: 1395417-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR