4,4,5,5-Tetramethyl-2-(2-methyl-4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Methyl-4-(trifluoromethyl)phenylboronic acid pinacol ester
31624-25G
31624-5G
76902-038EA
453.01
USD
76902-038
76902-040
4,4,5,5-Tetramethyl-2-(2-methyl-4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane ≥97%
4,4,5,5-Tetramethyl-2-(2-methyl-4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane
Formula:
C₁₄H₁₈BF₃O₂ MW: 286.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18729901 CAS Number: 1689509-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...