2-Fluoro-3-iodo-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

31549-1G 31549-25G 31549-500MG 31549-5G
76902-022EA 157.02 USD
76902-022 76902-024 76902-026 76902-028
2-Fluoro-3-iodo-4-methylbenzoic acid ≥97%
2-Fluoro-3-iodo-4-methylbenzoic acid
Formula: C₈H₆FIO₂
Storage Temperature: Ambient
CAS Number: 882679-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR