3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-formylphenylboronic acid pinacol ester

31438-1G 31438-25G 31438-500MG 31438-5G
76901-992EA 155.96 USD
76901-992 76901-994 76901-996 76901-998
3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥95%
3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Formula: C₁₃H₁₆BClO₃
MW: 266.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18712632
CAS Number: 1356642-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR