2-Bromo-5-methoxyisonicotinic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
31348-1G
31348-500MG
31348-5G
31348-25G
76901-962EA
498.63
USD
76901-962
76901-964
76901-966
77654-936
2-Bromo-5-methoxyisonicotinic acid ≥95%
2-Bromo-5-methoxyisonicotinic acid
Formula:
C₇H₆BrNO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD18257535 CAS Number: 1256789-55-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...