2-Chloro-4-fluoro-3-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

31242-1G 31242-25G 31242-500MG 31242-5G
76901-934EA 257.8 USD
76901-934 76901-936 76901-938 76901-940
2-Chloro-4-fluoro-3-methylbenzoic acid ≥95%
2-Chloro-4-fluoro-3-methylbenzoic acid
Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846444
CAS Number: 173315-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR