2-Chloro-3-fluorophenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

31138-1G 31138-25G 31138-5G
76901-908EA 62.6 USD
76901-908 76901-910 76901-912
2-Chloro-3-fluorophenylboronic acid pinacol ester ≥97% research grade
2-Chloro-3-fluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16996256
CAS Number: 1688698-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR