2,4-Difluoro-3-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31116-25G 31116-5G
76901-900EA 519.85 USD
76901-900 76901-902
2,4-Difluoro-3-formylphenylboronic acid pinacol ester ≥97%
2,4-Difluoro-3-formylphenylboronic acid pinacol ester
Formula: C₁₃H₁₅BF₂O₃
MW: 268.064 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088262
CAS Number: 2121512-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR