2-Chloro-5-methoxyphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

31072-25G 31072-5G
76901-866EA 456.19 USD
76901-866 76901-868
2-Chloro-5-methoxyphenylboronic acid pinacol ester ≥97% research grade
2-Chloro-5-methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₈BClO₃
Storage Temperature: Ambient
MDL Number: MFCD17215811
CAS Number: 1256781-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR