2,6-Dimethyl-4-cyanophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31032-1G 31032-500MG 31032-5G 31032-25G
76901-846EA 468.92 USD
76901-846 76901-848 76901-850 77658-620
2,6-Dimethyl-4-cyanophenylboronic acid pinacol ester ≥97%
2,6-Dimethyl-4-cyanophenylboronic acid pinacol ester
Formula: C₁₅H₂₀BNO₂
MW: 257.136 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24449621
CAS Number: 1448810-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR