2-Fluoro-5-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

31001-25G 31001-5G
76901-822EA 375.56 USD
76901-822 76901-824
2-Fluoro-5-methoxyphenylboronic acid pinacol ester ≥97%
2-Fluoro-5-methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₈BFO₃
MDL Number: MFCD18384134
CAS Number: 1190129-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR