2-Bromo-4-fluoro-3-methyl-1-(methylthio)-benzene ≥97%

Supplier: AOBChem USA

Warning

30959-1G 30959-25G 30959-500MG 30959-5G
76901-804EA 252.5 USD
76901-804 76901-806 76901-808 76901-810
2-Bromo-4-fluoro-3-methyl-1-(methylthio)-benzene ≥97%
2-Bromo-4-fluoro-3-methyl-1-(methylthio)-benzene
Formula: C₈H₈BrFS
MW: 235.117 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088240
CAS Number: 1879026-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR