(2-Chloro-3,4-difluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-3,4-difluorobenzyl alcohol

30939-1G 30939-25G 30939-500MG 30939-5G
76901-788EA 129.43 USD
76901-788 76901-790 76901-792 76901-794
(2-Chloro-3,4-difluorophenyl)methanol ≥97%
(2-Chloro-3,4-difluorophenyl)methanol
Formula: C₇H₅ClF₂O
MW: 178.56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28520354
CAS Number: 1784873-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR