2,6-Dimethoxy-5-methylpyridine-3-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

30755-1G 30755-25G 30755-500MG 30755-5G
76901-744EA 231.28 USD
76901-744 76901-746 76901-748 76901-750
2,6-Dimethoxy-5-methylpyridine-3-carboxaldehyde ≥97%
2,6-Dimethoxy-5-methylpyridine-3-carboxaldehyde
Formula: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD29088196
CAS Number: 1826110-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR