2,6-Dimethyl-4-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

30723-1G 30723-25G 30723-500MG 30723-5G
76901-730EA 231.28 USD
76901-730 76901-732 76901-734 76901-736
2,6-Dimethyl-4-fluorobenzyl alcohol ≥97%
2,6-Dimethyl-4-fluorobenzyl alcohol
Formula: C₉H₁₁FO
Storage Temperature: Refrigerator
MDL Number: MFCD03701056
CAS Number: 773868-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR