2-Chloro-5-methoxy-3-pyridinecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

30710-1G 30710-25G 30710-500MG 30710-5G
76901-702EA 260.99 USD
76901-702 76901-704 76901-706 76901-708
2-Chloro-5-methoxy-3-pyridinecarboxylic acid ≥97%
2-Chloro-5-methoxy-3-pyridinecarboxylic acid
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16610607
CAS Number: 74650-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR