2-Chloro-4-fluoro-6-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

30685-1G 30685-25G 30685-500MG 30685-5G
76901-690EA 409.51 USD
76901-690 76901-692 76901-694 76901-696
2-Chloro-4-fluoro-6-methylbenzaldehyde ≥97%
2-Chloro-4-fluoro-6-methylbenzaldehyde
Formula: C₈H₆ClFO
MW: 172.584 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28751751
CAS Number: 1805524-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR