2-Benzyloxy-5-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2-(Benzyloxy)-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

30661-1G 30661-25G 30661-500MG 30661-5G
76901-666EA 225.98 USD
76901-666 76901-668 76901-670 76901-672
2-Benzyloxy-5-methylphenylboronic acid pinacol ester ≥97%
2-Benzyloxy-5-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₅BO₃
MW: 324.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13152011
CAS Number: 1204580-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR