2-Bromo-4-(tert-butyl)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

30637-1G 30637-25G 30637-5G
76901-652EA 114.58 USD
76901-652 76901-654 76901-656
2-Bromo-4-(tert-butyl)-1-methylbenzene ≥97%
2-Bromo-4-(tert-butyl)-1-methylbenzene
Formula: C₁₁H₁₅Br
Storage Temperature: Ambient
MDL Number: MFCD00154985
CAS Number: 61024-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR