2-Bromo-6-fluoro-3'-methylbiphenyl ≥95%

Supplier: AOBChem USA

Warning

30590-1G 30590-25G 30590-500MG 30590-5G
76901-630EA 337.37 USD
76901-630 76901-632 76901-634 76901-636
2-Bromo-6-fluoro-3'-methylbiphenyl ≥95%
2-Bromo-6-fluoro-3'-methylbiphenyl
Formula: C₁₃H₁₀BrF
Storage Temperature: Refrigerator
MDL Number: MFCD29088167
CAS Number: 884512-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR