2-Benzyloxy-3-pyridinecarbaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

30534-1G 30534-25G 30534-500MG 30534-5G
76901-584EA 185.66 USD
76901-584 76901-586 76901-588 76901-590
2-Benzyloxy-3-pyridinecarbaldehyde ≥97% research grade
2-Benzyloxy-3-pyridinecarbaldehyde
Formula: C₁₃H₁₁NO₂
MW: 213.232 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18256123
CAS Number: 179257-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR