2,6-Dichlorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30510-25G 30510-5G
76901-574EA 240.83 USD
76901-574 76901-576
2,6-Dichlorophenylboronic acid pinacol ester ≥97%
2,6-Dichlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BCl₂O₂
MW: 272.963 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18729914
CAS Number: 69807-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR