2,6-Difluoro-4-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,3-Difluoro-5-iodo-2-methoxybenzene

30433-25G 30433-5G
76901-548EA 346.92 USD
76901-548 76901-550
2,6-Difluoro-4-iodoanisole ≥97%
2,6-Difluoro-4-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
MW: 270.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368787
CAS Number: 886762-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR