2-Fluoro-4-methoxybenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2-Fluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-4-methoxyphenylboronic acid pinacol ester

30430-25G 30430-5G
76901-540EA 304.48 USD
76901-540 76901-542
2-Fluoro-4-methoxybenzeneboronic acid pinacol ester ≥97%
2-Fluoro-4-methoxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16996299
CAS Number: 628692-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR