2,6-Difluoro-3-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

30425-1G 30425-25G 30425-500MG 30425-5G
76901-528EA 260.99 USD
76901-528 76901-530 76901-532 76901-534
2,6-Difluoro-3-iodobenzaldehyde ≥97%
2,6-Difluoro-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₂IO
Storage Temperature: Ambient
MDL Number: MFCD11845973
CAS Number: 1160573-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR