2,6-Difluoro-4-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoropyridine-4-boronic acid pinacol ester, 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

30410-1G 30410-25G 30410-500MG 30410-5G
76901-512EA 206.88 USD
76901-512 76901-514 76901-516 76901-518
2,6-Difluoro-4-pyridineboronic acid pinacol ester ≥97%
2,6-Difluoro-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BF₂NO₂
MW: 241.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913987
CAS Number: 1204333-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR