(2-Methoxy-6-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-6-methylphenylboronic acid

30265-1G 30265-25G 30265-500MG 30265-5G
76901-476EA 263.11 USD
76901-476 76901-478 76901-480 76901-482
(2-Methoxy-6-methylphenyl)boronic acid ≥97%
(2-Methoxy-6-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD11043444
CAS Number: 1567218-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR