2-Hydroxy-5-(trifluoromethyl)benzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
30241-1G
30241-25G
30241-5G
76901-464EA
81.69
USD
76901-464
76901-466
76901-468
2-Hydroxy-5-(trifluoromethyl)benzaldehyde ≥97%
2-Hydroxy-5-(trifluoromethyl)benzaldehyde
Formula:
C₈H₅F₃O₂ MW: 190.11 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16999709 CAS Number: 210039-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...