2-Formyl-4-(trifluoromethyl)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

30211-1G 30211-25G 30211-500MG 30211-5G
76901-448EA 409.51 USD
76901-448 76901-450 76901-452 76901-454
2-Formyl-4-(trifluoromethyl)benzoic acid ≥97% research grade
2-Formyl-4-(trifluoromethyl)benzoic acid
Formula: C₉H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD22042204
CAS Number: 23984-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR