2-Chloro-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine

30029-1G 30029-25G 30029-500MG 30029-5G
76901-406EA 260.99 USD
76901-406 76901-408 76901-410 76901-412
2-Chloro-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester ≥97%
2-Chloro-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClF₃NO₂
MW: 307.5000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195760
CAS Number: 1218790-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR