2-Bromo-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29819-1G 29819-25G 29819-5G
76901-338EA 62.6 USD
76901-338 76901-340 76901-342
2-Bromo-3-methylbenzaldehyde ≥97%
2-Bromo-3-methylbenzaldehyde
Formula: C₈H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD18392002
CAS Number: 109179-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR