2-Bromo-4,5-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-3,4-xylenol

29587-1G 29587-25G 29587-5G
76901-316EA 114.58 USD
76901-316 76901-318 76901-320
2-Bromo-4,5-dimethylphenol ≥97%
2-Bromo-4,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09260834
CAS Number: 22802-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR