(2-Bromo-3,4-difluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-3,4-difluorobenzyl alcohol

29584-1G 29584-25G 29584-500MG 29584-5G
76901-304EA 132.62 USD
76901-304 76901-306 76901-308 76901-310
(2-Bromo-3,4-difluorophenyl)methanol ≥97%
(2-Bromo-3,4-difluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.015 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28657913
CAS Number: 1578156-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR