2-Chloro-3,5-difluorobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

29502-1G 29502-25G 29502-5G
76901-284EA 75.33 USD
76901-284 76901-286 76901-288
2-Chloro-3,5-difluorobenzoic acid ≥97% research grade
2-Chloro-3,5-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18415557
CAS Number: 189024-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR