2-Chloro-4,5-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

29473-1G 29473-25G 29473-500MG 29473-5G
76901-252EA 292.81 USD
76901-252 76901-254 76901-256 76901-258
2-Chloro-4,5-difluorophenylboronic acid ≥97%
2-Chloro-4,5-difluorophenylboronic acid
Formula: C₆H₄BClF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12911677
CAS Number: 1801916-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR