2,5-Dibromopyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

29386-1G 29386-25G 29386-500MG 29386-5G
76901-236EA 290.69 USD
76901-236 76901-238 76901-240 76901-242
2,5-Dibromopyridin-3-ol ≥97%
2,5-Dibromopyridin-3-ol
Formula: C₅H₃Br₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD09959722
CAS Number: 857429-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR