2-Fluoro-6-methoxy-4-methyl-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

29320-1G 29320-25G 29320-500MG 29320-5G
76901-222EA 142.17 USD
76901-222 76901-224 76901-226 76901-228
2-Fluoro-6-methoxy-4-methyl-benzoic acid ≥97%
2-Fluoro-6-methoxy-4-methyl-benzoic acid
Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712070
CAS Number: 1427416-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR