2-Chloro-6-iodo-3-pyridinol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-6-iodopyridin-3-ol

29303-25G 29303-5G
76901-200EA 268.41 USD
76901-200 76901-202
2-Chloro-6-iodo-3-pyridinol ≥97%
2-Chloro-6-iodo-3-pyridinol
Formula: C₅H₃ClINO
MW: 255.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566651
CAS Number: 185220-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR